Invitrogen
Manufacturer Code:PA316805
Catalog # APSA6805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Extracellular residues 230-380 of Scavenger Receptor-BI/BII expressed as two tandem copies in bacteria using the pET system. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 85 kDa lysosomal membrane sialoglycoprotein; 85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2 (lysosomal integral membrane protein II); CD36 antigen-like 1; CD36 antigen-like 2; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; HDL QTL 1; LGP85; LIMP II; Lysosome membrane protein 2; Lysosome membrane protein II; Scarb1 Scarb2 SR-B1/B2 SR-BI/BII Scavenger Receptor-BI/BII Scavenger Receptor-B1/B2 Scavenger Receptor B1 Scavenger Receptor B2; scavanger receptor class B type 1; Scavenger receptor class B member 1; Scavenger receptor class B member 2; scavenger receptor class B type I; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B, member 2; scavenger receptor class B1; SR-BI; SRB1
Gene Aliases: 9330185J12Rik; AI120173; AMRF; CD36; CD36L1; CD36L2; CLA-1; CLA1; D5Ertd460e; EPM4; Hdlq1; HDLQTL6; Hlb398; HLGP85; LGP85; LIMP-2; LIMP2; LIMPII; MLGP85; mSR-BI; SCARB1; SCARB2; SR-B1; SR-BI; SR-BII; SRB1; SRBI
UniProt ID: (Human) Q8WTV0, (Human) Q14108, (Rat) P97943, (Mouse) Q61009, (Rat) P27615, (Mouse) Q3UNF8
Entrez Gene ID: (Human) 949, (Human) 950, (Rat) 25073, (Mouse) 20778, (Rat) 117106, (Mouse) 12492
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.