R&D Systems
Manufacturer Code:MAB8114SP
Catalog # MAB8114SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 947007 |
Immunogen | Human embryonic kidney cell line HEK293-derived human SR-BI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD36; CD36 and LIMPII analogous 1; CD36 and LIMPII analogous 1 CD36 antigen CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 CD36L1 CD36L1scavenger receptor class B type 1 CLA1 CLA1 CD36 antigen-like 1 CLA-1 HDLQTL6 Collagen type I receptor thrombospondin receptor-like 1 HDLQTL6 MGC138242 SCARB1 scavenger receptor class B type III scavenger receptor class B member 1 SR-B1 SRB1 scavenger receptor class B member 1 SRBI; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen-like 1; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; Scavenger receptor class B member 1; scavenger receptor class B type III; scavenger receptor class B, member 1; SR-BI; SRB1
Gene Aliases: CD36L1; CLA-1; CLA1; HDLQTL6; SCARB1; SR-BI; SRB1
UniProt ID: (Human) Q8WTV0
Entrez Gene ID: (Human) 949
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.