Novus Biologicals
Manufacturer Code:NB400131SS
Catalog # NB400131SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the C-terminal region of mouse SR-BI (between residues 400-509). [UniProt# Q61009] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD36; CD36 and LIMPII analogous 1; CD36 and LIMPII analogous 1 CD36 antigen CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 CD36L1scavenger receptor class B type 1 CLA1 CD36 antigen-like 1 CLA-1 HDLQTL6 Collagen type I receptor thrombospondin receptor-like 1 MGC138242 SCARB1 scavenger receptor class B type III scavenger receptor class B member 1 SR-B1 SRB1 scavenger receptor class B member 1 SRBI; CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen-like 1; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; Scavenger receptor class B member 1; scavenger receptor class B type III; scavenger receptor class B, member 1; SR-BI; SRB1
Gene Aliases: CD36L1; CLA-1; CLA1; HDLQTL6; SCARB1; SR-BI; SRB1
UniProt ID: (Human) Q8WTV0
Entrez Gene ID: (Human) 949
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.