Invitrogen
Manufacturer Code:PA116803
Catalog # APPA6803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the C-terminal residues 400-509 of mouse SR-BI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen-like 1; CD36-related class B scavenger receptor haSR-BI; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; HaSR-BI; HDL QTL 1; SCARB1 SR-B1 SR-BI Scavenger Receptor-BI Scavenger Receptor-B1; scavanger receptor class B type 1; Scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SR-BI; SRB1
Gene Aliases: AI120173; CD36; CD36L1; CLA-1; CLA1; D5Ertd460e; haSR-bI; Hdlq1; HDLQTL6; Hlb398; mSR-BI; SCARB1; SR-B1; SR-BI; SRB1; SRBI
UniProt ID: (Human) Q8WTV0, (Rat) P97943, (Mouse) Q61009, (Chinese hamster) Q60417
Entrez Gene ID: (Human) 949, (Rat) 25073, (Mouse) 20778, (Chinese hamster) 100736552
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.