Proteintech
Manufacturer Code:172561AP150UL
Catalog # 50-557-193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SQRDL Fusion Protein Ag10762 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 44 SQRDL; flavo-binding protein; SQOR; sulfide dehydrogenase like; Sulfide dehydrogenase-like; Sulfide quinone oxidoreductase; Sulfide:quinone oxidoreductase, mitochondrial
Gene Aliases: 0610039J17Rik; 4930557M22Rik; Cc2-17; CGI-44; PRO1975; SQOR; SQRDL
UniProt ID: (Human) Q9Y6N5, (Mouse) Q9R112
Entrez Gene ID: (Human) 58472, (Rat) 691966, (Mouse) 59010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.