Invitrogen
Manufacturer Code:PA598858
Catalog # PIPA598858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Serine palmitoyltransferase 3 protein (1-58AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LCB 3; LCB2b; Long chain base biosynthesis protein 2b; Long chain base biosynthesis protein 3; Serine palmitoyltransferase 3; serine palmitoyltransferase, long chain base subunit 2-like (aminotransferase 2); serine palmitoyltransferase, long chain base subunit 3; Serine-palmitoyl-CoA transferase 3; Serine-palmityl-CoA transferase 3; SPT 3
Gene Aliases: C130053K05Rik; C20orf38; dJ718P11; dJ718P11.1; hLCB2b; LCB2B; LCB3; SPT3; SPTLC2L; SPTLC3
UniProt ID: (Human) Q9NUV7, (Mouse) Q8BG54
Entrez Gene ID: (Human) 55304, (Mouse) 228677
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.