Invitrogen
Manufacturer Code:PA565493
Catalog # PIPA565493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human SPTLC3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf38 SPTLC2L; LCB 3; LCB2b; Long chain base biosynthesis protein 2b; Long chain base biosynthesis protein 3; Serine palmitoyltransferase 3; serine palmitoyltransferase, long chain base subunit 2-like (aminotransferase 2); serine palmitoyltransferase, long chain base subunit 3; Serine-palmitoyl-CoA transferase 3; SPT 3
Gene Aliases: C20orf38; dJ718P11; dJ718P11.1; hLCB2b; LCB2B; LCB3; SPT3; SPTLC2L; SPTLC3
UniProt ID: (Human) Q9NUV7
Entrez Gene ID: (Human) 55304
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.