Invitrogen
Manufacturer Code:PA531130
Catalog # PIPA531130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 50 and 274 of Human Serine palmitoyltransferase 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSN1C LCB2 LCB2A NSAN1C SPT2 hLCB2a LCB 2 SPT 2 long chain base biosynthesis protein 2a serine palmitoyltransferase 2 serine palmitoyltransferase subunit II serine-palmitoyl-CoA transferase 2; LCB 2; LCB2a; Long chain base biosynthesis protein 2; Long chain base biosynthesis protein 2a; Serine palmitoyltransferase 2; serine palmitoyltransferase, long chain base subunit 2; serine palmitoyltransferase, subunit II; Serine-palmitoyl-CoA transferase 2; SPT 2
Gene Aliases: AI173915; hLCB2a; HSN1C; KIAA0526; LCB2; LCB2A; mKIAA0526; NSAN1C; SPT2; SPTLC2
UniProt ID: (Human) O15270, (Mouse) P97363
Entrez Gene ID: (Human) 9517, (Mouse) 20773
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.