Invitrogen
Manufacturer Code:PA521142
Catalog # PIPA521142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the carboxy terminus of human SPT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSN1C LCB2 LCB2A NSAN1C SPT2 hLCB2a LCB 2 SPT 2 long chain base biosynthesis protein 2a serine palmitoyltransferase 2 serine palmitoyltransferase subunit II serine-palmitoyl-CoA transferase 2; LCB 2; LCB2a; Long chain base biosynthesis protein 2; Long chain base biosynthesis protein 2a; protein-O-mannosyltransferase 2; Serine palmitoyltransferase 2; serine palmitoyltransferase, long chain base subunit 2; serine palmitoyltransferase, subunit II; Serine-palmitoyl-CoA transferase 2; SPT 2
Gene Aliases: AI173915; hLCB2a; HSN1C; KIAA0526; LCB2; LCB2A; mKIAA0526; NSAN1C; Pomt2; SPT2; SPTLC2
UniProt ID: (Human) O15270, (Mouse) P97363, (Rat) Q3B7D2
Entrez Gene ID: (Human) 9517, (Mouse) 20773, (Rat) 366697
Molecular Function: transaminase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.