Invitrogen
Manufacturer Code:PA542914
Catalog # PIPA542914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SPTLC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LCB 1; Long chain base biosynthesis protein 1; serine C-palmitoyltransferase; Serine palmitoyltransferase 1; Serine palmitoyltransferase long chain base subunit 1 HSAN HSAN1 HSN1 LBC1 LCB1 MGC14645 SPT1 SPTI hLCB1; serine palmitoyltransferase subunit 1; serine palmitoyltransferase, long chain base subunit 1; Serine-palmitoyl-CoA transferase 1; SPT 1; SPT1
Gene Aliases: AW552086; C77762; E030036H05; fc75h04; fp41c08; HSAN1; HSN1; LBC1; LCB1; rCG_44191; RGD1306617; SPT1; SPTI; SPTLC1; wu:fc75h04; wu:fp41c08; zgc:112247
UniProt ID: (Bovine) Q3MHG1, (Human) O15269, (Mouse) O35704, (Rat) D4A2H2
Entrez Gene ID: (Bovine) 614165, (Horse) 100063025, (Dog) 484192, (Rabbit) 100344536, (Human) 10558, (Guinea pig) 100726468, (Mouse) 268656, (Rat) 361213, (Zebrafish) 553981
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.