Abnova
Manufacturer Code:PAB19392
Catalog # 89-120-928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids near C-terminus of human SPTLC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SPTLC1.
For Research Use Only
Protein Aliases: HSAN; HSAN1; HSN1; LBC1; LCB 1; LCB1; Long chain base biosynthesis protein 1; MGC14645; serine C-palmitoyltransferase; Serine palmitoyltransferase 1; serine palmitoyltransferase, long chain base subunit 1; Serine-palmitoyl-CoA transferase 1; SPT 1; SPT1; SPTI
Gene Aliases: HSAN1; HSN1; LBC1; LCB1; SPT1; SPTI; SPTLC1
UniProt ID: (Human) O15269
Entrez Gene ID: (Human) 10558
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.