Abnova
Manufacturer Code:MAB14992
Catalog # 89-035-012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SPTBN2/1582 |
Immunogen | Recombinant protein corresponding to amino acids 356-475 of human SPTBN2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-III spectrin; glutamate transporter EAAT4-associated protein 41; GTRAP41; SCA5; spectrin beta chain, brain 2; Spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; Spinocerebellar ataxia 5 protein
Gene Aliases: GTRAP41; KIAA0302; SCA5; SCAR14; SPTBN2
UniProt ID: (Human) O15020
Entrez Gene ID: (Human) 6712
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.