Proteintech
Manufacturer Code:197221AP150UL
Catalog # 50-173-2508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta fodrin; Beta II spectrin beta-fodrin betaSpII ELF Fodrin beta chain Spectrin beta chain brain 1 SPTB2 SPTBN1; beta-fodrin; beta-G spectrin; Beta-II spectrin; beta-spectrin 2; beta-spectrin 2, non-erythrocytic; beta-spectrin II; beta-spectrin non-erythrocytic 1; brain spectrin; embryonic liver beta-fodrin; Embryonic liver fodrin; epididymis luminal protein 102; Fodrin beta chain; non-erythrocytic; spectrin beta 2; spectrin beta chain, brain 1; Spectrin beta chain, non-erythrocytic 1; spectrin G; spectrin, beta, non-erythrocytic 1; Spectrin, non-erythroid beta chain 1
Gene Aliases: 9930031C03Rik; AL033301; betaSpII; ELF; elf1; elf3; HEL102; mKIAA4049; Spnb-2; Spnb2; SPTB2; SPTBN1
UniProt ID: (Human) Q01082, (Mouse) Q62261
Entrez Gene ID: (Human) 6711, (Mouse) 20742
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.