Abnova
Manufacturer Code:MAB2508
Catalog # 89-111-043
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Chicken |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AA6 |
Immunogen | Native purified chicken SPTAN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Mouse monoclonal antibody raised against native SPTAN1.
For Research Use Only
Protein Aliases: Alpha-II spectrin; alpha-spectrin; Fodrin alpha chain; nonerythroid alpha-spectrin; SPECA; spectrin alpha chain, brain; Spectrin alpha chain, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1 (alpha-fodrin); spectrin, non-erythroid alpha chain
Gene Aliases: SPECA; SPTA2; SPTAN1
UniProt ID: (Chicken) P07751
Entrez Gene ID: (Chicken) 374234
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.