Invitrogen
Manufacturer Code:PA540709
Catalog # PIPA540709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SUPT5H |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DRB sensitivity-inducing factor 160 kDa subunit; DRB sensitivity-inducing factor large subunit; DSIF large subunit; DSIF p160; hSPT5; protein foggy; suppressor of Ty 5 homolog; Suppressor of Ty 5 homolog (S. cerevisiae) SPT5 SPT5H Tat-CT1; Tat-cotransactivator 1 protein; Tat-CT1 protein; Transcription elongation factor SPT5; transcription elongation regulator foggy
Gene Aliases: etID10129.15; etID37101.15; fb37g01; fb95e05; fog; foggy; SPT5; SPT5H; SUPT5H; Tat-CT1; wu:fb37g01; wu:fb95e05; wu:fc10f02
UniProt ID: (Human) O00267
Entrez Gene ID: (Human) 6829, (Dog) 476462, (Horse) 100064516, (Bovine) 519808, (Rabbit) 100356793, (Guinea pig) 100713914, (Zebrafish) 64275
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.