Novus Biologicals
Manufacturer Code:NBP274348AF700
Catalog # N274348A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A5 |
Immunogen | Full length recombinant protein of human SPT3 (NP_852001) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: SPT3-like protein; SPT3-like protein SPT3SPT3L suppressor of Ty (S.cerevisiae) 3 homolog suppressor of Ty 3 homolog (S. cerevisiae) transcription initiation protein SPT3 homolog; suppressor of Ty 3 homolog; Transcription initiation protein SPT3 homolog
Gene Aliases: SPT3; SPT3L; SUPT3H
UniProt ID: (Human) A6NKG9
Entrez Gene ID: (Human) 8464
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.