Invitrogen
Manufacturer Code:PA540315
Catalog # PIPA540315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SUPT3H |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cdc5-like protein; SPT3-like protein; suppressor of Ty 3 homolog; suppressor of Ty 3 homolog (S. cerevisiae); Suppressor of Ty 3 homolog (S. cerevisiae) SPT3 SPT3L; Transcription initiation protein SPT3 homolog
Gene Aliases: SPT3; SPT3L; SUPT3H
UniProt ID: (Human) A6NKG9
Entrez Gene ID: (Dog) 474922, (Horse) 100068070, (Bovine) 618133, (Human) 8464, (Goat) 100861008, (Rabbit) 100338662, (Pig) 100154712, (Rat) 685697, (Guinea pig) 100734454
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.