Abnova
Manufacturer Code:PAB1163
Catalog # 89-115-235
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 1-200 <i>Saccharomyces cerevisiae</i> SPT3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyoclonal antibody raised against partial recombinant Saccharomyces cerevisiae SPT3.
For Research Use Only
Protein Aliases: Positive regulator of Ty transcription; Protein SPT3; transcriptional regulator SPT3
Gene Aliases: D9509.12; SPT3; YDR392W
UniProt ID: (Baker's yeast) P06844
Entrez Gene ID: (Baker's yeast) 852001
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.