Invitrogen
Manufacturer Code:PA521170
Catalog # PIPA521170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the carboxy terminus of human SPRYD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310076E16Rik; C5orf10 SPRYD2 TRIM76 2310076E16Rik SPRY domain-containing protein 2 cardiomyopathy-associated protein 5 dystrobrevin-binding protein 2 genethonin-3 myospryn tripartite motif-containing 76 tripartite motif-containing protein 76; cardiomyopathy associated 5; Cardiomyopathy-associated protein 5; Dystrobrevin-binding protein 2; Genethonin-3; heart and skeletal muscle-specific and sprouty domain-containing; Myospryn; SPRY domain-containing protein 2; stretch response protein 553; stretch-response protein 553; stretch-responsive fibronectin protein type 3; TRIM-like protein; tripartite motif-containing 76; Tripartite motif-containing protein 76
Gene Aliases: 2310076E16Rik; 2310076E21Rik; AA420382; AI504003; C5orf10; CMYA5; DTNBP2; SPRYD2; sr553; Srfsd; Tims; TRIM76
UniProt ID: (Human) Q8N3K9
Entrez Gene ID: (Human) 202333, (Rat) 688915, (Mouse) 76469
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.