Invitrogen
Manufacturer Code:PA543584
Catalog # PIPA543584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SPRY2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein sprouty homolog 2; sprouty 2; sprouty homolog 2; Sprouty homolog 2 (Drosophila) MGC23039 hSPRY2; Spry-2
Gene Aliases: hSPRY2; IGAN3; sprouty2; SPRY2
UniProt ID: (Bovine) Q08E39, (Human) O43597, (Mouse) Q9QXV8
Entrez Gene ID: (Dog) 485504, (Pig) 100157266, (Rabbit) 100352158, (Bovine) 539090, (Human) 10253, (Horse) 100050086, (Mouse) 24064, (Rat) 306141, (Guinea pig) 100729578
Molecular Function: signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.