Novus Biologicals
Manufacturer Code:NBP204100
Catalog # NBP204100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 265 and 315 of human Sprouty 2 using the numbering given in entry NP_005833.1 (GeneID 10253). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSPRY2 MGC23039 protein sprouty homolog 2 sprouty (Drosophila) homolog 2 sprouty 2 sprouty homolog 2 (Drosophila) spry-2; Protein sprouty homolog 2; Spry-2
Gene Aliases: hSPRY2; IGAN3; SPRY2
UniProt ID: (Human) O43597
Entrez Gene ID: (Human) 10253
Molecular Function:
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.