Abnova
Manufacturer Code:H00010253A01
Catalog # 89-106-253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. &bsim 315 a.a) full-length recombinant protein with GST tag.,SPRY2 (AAH15745.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant SPRY2.
For Research Use Only
Protein Aliases: hSPRY2; MGC23039; Protein sprouty homolog 2; Spry-2
Gene Aliases: hSPRY2; IGAN3; SPRY2
UniProt ID: (Human) O43597
Entrez Gene ID: (Human) 10253
Molecular Function: signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.