Proteintech
Manufacturer Code:113831AP150UL
Catalog # 50-172-7112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPRY2 Fusion Protein Ag1918 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSPRY2 Protein sprouty homolog 2 sprouty homolog 2 (Drosophila) Spry 2 SPRY2; Protein sprouty homolog 2; sprouty 2; Spry-2
Gene Aliases: hSPRY2; IGAN3; sprouty2; SPRY2
UniProt ID: (Human) O43597, (Mouse) Q9QXV8
Entrez Gene ID: (Human) 10253, (Mouse) 24064, (Rat) 306141
Molecular Function:
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.