R&D Systems
Manufacturer Code:MAB6097
Catalog # MAB6097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 669534 |
Immunogen | <EM>E.coli</EM>-derived recombinant human SPRY1 Met1-Lys178 Accession # O43609 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Drosophila homolog of 1 (antagonist of FGF signaling) sprouty homolog 1 antagonist of FGF signaling (Drosophila) spry-1; Protein sprouty homolog 1; sprouty homolog 1, antagonist of FGF signaling; sprouty, Drosophila, homolog of, 1 (antagonist of FGF signaling); Spry-1
Gene Aliases: hSPRY1; SPRY1
UniProt ID: (Human) O43609
Entrez Gene ID: (Human) 10252
Molecular Function: signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.