Invitrogen
Manufacturer Code:PA543582
Catalog # PIPA543582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SPRY1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein sprouty homolog 1; sprouty 1; Sprouty homolog 1 antagonist of FGF signaling (Drosophila) hSPRY1; sprouty homolog 1, antagonist of FGF signaling; sprouty, Drosophila, homolog of, 1 (antagonist of FGF signaling); sprouty1; Spry-1
Gene Aliases: hSPRY1; sprouty1; spry-1; SPRY1
UniProt ID: (Bovine) A5D992, (Human) O43609, (Mouse) Q9QXV9
Entrez Gene ID: (Rabbit) 100357926, (Dog) 483838, (Bovine) 507095, (Human) 10252, (Horse) 100063739, (Mouse) 24063, (Rat) 294981, (Guinea pig) 100734287, (Zebrafish) 798229
Molecular Function:
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.