Invitrogen
Manufacturer Code:PA588215
Catalog # PIPA588215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-261 of human SPR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sepiapterin reductase; short chain dehydrogenase/reductase family 38C, member 1; SPR
Gene Aliases: AA409688; ENSMUSG00000071345; Gm10328; SDR38C1; SPR
UniProt ID: (Human) P35270, (Rat) P18297
Entrez Gene ID: (Human) 6697, (Mouse) 20751, (Rat) 29270
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.