Invitrogen
Manufacturer Code:PA542683
Catalog # PIPA542683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SPPL2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IMP-4; Intramembrane protease 4; Presenilin homologous protein 4; Presenilin-like protein 1; protein SPP-like 2B; protein SPPL2b; PSH4; Signal peptide peptidase like 2B IMP4 KIAA1532 MGC111084 PSL1 IMP-4; Signal peptide peptidase-like 2B; SPP-like 2B
Gene Aliases: 3110056O03Rik; AW550292; IMP-4; IMP4; KIAA1532; PSH4; PSL1; RGD1308556; SPPL2B
UniProt ID: (Human) Q8TCT7, (Rat) Q5PQL3, (Mouse) Q3TD49
Entrez Gene ID: (Dog) 485071, (Horse) 100060546, (Human) 56928, (Pig) 100515648, (Guinea pig) 100719910, (Rat) 362828, (Mouse) 73218
Molecular Function: aspartic protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.