Abnova
Manufacturer Code:PAB4907
Catalog # 89-131-639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human SPPL2A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SPPL2A.
For Research Use Only
Protein Aliases: IMP-3; IMP3; intramembrane cleaving protease; Intramembrane protease 3; Presenilin-like protein 2; PSL2; Signal peptide peptidase-like 2A; SPP-like 2A
Gene Aliases: IMP3; PSEC0147; PSL2; SPPL2A
UniProt ID: (Human) Q8TCT8
Entrez Gene ID: (Human) 84888
Molecular Function: aspartic protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.