Proteintech
Manufacturer Code:132551AP150UL
Catalog # 50-172-8606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPPL2A Fusion Protein Ag3935 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IMP 3 IMP3 Intramembrane protease 3 Presenilin like protein 2 PSL2 SPP like 2A SPPL2A; IMP-3; intramembrane cleaving protease; Intramembrane protease 3; Presenilin-like protein 2; protein SPPL2a; Signal peptide peptidase-like 2A; SPP-like 2A; SPP-like 2A protein
Gene Aliases: 2010106G01Rik; C130089K23Rik; IMP3; MNCb-3763; PSEC0147; PSL2; RGD1563001; SPPL2A
UniProt ID: (Human) Q8TCT8, (Mouse) Q9JJF9
Entrez Gene ID: (Human) 84888, (Rat) 311401, (Mouse) 66552
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.