Proteintech
Manufacturer Code:204161AP150UL
Catalog # 50-173-2877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPP Fusion Protein Ag14158 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H13 hIMP1 histocompatibility (minor) 13 HM13 IMP 1 IMP1 IMPAS IMPAS 1 Intramembrane protease 1 MSTP086 Presenilin like protein 3 PSENL3 PSL3 Signal peptide peptidase SPP; histocompatibility (minor) 13; histocompatibility 13; IMP-1; Intramembrane protease 1; minor histocompatibility antigen 13; Minor histocompatibility antigen H13; presenilin-like aspartyl protease; Presenilin-like protein 3; Signal peptide peptidase; signal peptide peptidase beta; signal peptide peptidase like 1
Gene Aliases: 1200006O09Rik; 4930443L17Rik; 5031424B04Rik; AV020344; H-13; H13; HM13; IMP1; IMPAS; IMPAS-1; MSTP086; PSENL3; PSL3; SPP; SPPL1
UniProt ID: (Mouse) Q9D8V0, (Human) Q8TCT9
Entrez Gene ID: (Mouse) 14950, (Human) 81502
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.