Proteintech
Manufacturer Code:167501AP150UL
Catalog # 50-560-729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPOP Fusion Protein Ag10215 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HIB homolog 1; HIB homolog 1 Roadkill homolog 1 speckle type POZ protein SPOP TEF2; PDX-1 C-terminal-interacting factor 1; Roadkill homolog 1; speckle type POZ protein; Speckle-type POZ protein
Gene Aliases: AI315626; BTBD32; Pcif1; SPOP; TEF2
UniProt ID: (Human) O43791, (Mouse) Q6ZWS8
Entrez Gene ID: (Human) 8405, (Mouse) 20747, (Rat) 287643
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
hydrolase
non-motor actin binding protein
protease
serine protease
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.