Proteintech
Manufacturer Code:205131AP150UL
Catalog # 50-173-2929
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPON2 Fusion Protein Ag14337 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Differentially expressed in cancerous and non-cancerous lung cells 1; DIL 1 DIL1 M spondin Mindin SPON2 Spondin 2; DIL-1; Mindin; spondin 2, extracellular matrix protein; Spondin-2
Gene Aliases: DIL-1; DIL1; M-SPONDIN; MINDIN; SPON2; UNQ435/PRO866
UniProt ID: (Human) Q9BUD6
Entrez Gene ID: (Human) 10417
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.