Invitrogen
Manufacturer Code:PA544237
Catalog # PIPA544237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SPO11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 35; CT35; Meiotic recombination protein SPO11; spermatogenesis associated 43; SPO11 meiotic protein covalently bound to DSB homolog; SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae) MGC39953 CT35; sporulation protein, meiosis-specific, SPO11 homolog; sporulation protein, meiosis-specific, SPO11 homolog (S. cervisiae)
Gene Aliases: AI449549; CT35; SPATA43; SPO11; TOPVIA; zgc:77876
UniProt ID: (Human) Q9Y5K1, (Mouse) Q9WTK8
Entrez Gene ID: (Dog) 610247, (Bovine) 524105, (Rabbit) 100359270, (Human) 23626, (Horse) 100050365, (Mouse) 26972, (Rat) 366261, (Guinea pig) 100725083, (Zebrafish) 402982
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.