Proteintech
Manufacturer Code:155091AP150UL
Catalog # 50-173-0278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPO11 Fusion Protein Ag4340 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 35; Cancer/testis antigen 35 CT35 SPO11; CT35; Meiotic recombination protein SPO11; spermatogenesis associated 43; SPO11 meiotic protein covalently bound to DSB homolog; sporulation protein, meiosis-specific, SPO11 homolog (S. cervisiae)
Gene Aliases: AI449549; CT35; SPATA43; SPO11; TOPVIA
UniProt ID: (Human) Q9Y5K1, (Mouse) Q9WTK8
Entrez Gene ID: (Human) 23626, (Mouse) 26972
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.