Novus Biologicals
Manufacturer Code:NBP179813
Catalog # NBP179813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human SPIRE2The immunogen for this antibody is SPIRE2. Peptide sequence EAQTVQSLGFAIYRALDWGLDESEERELSPQLERLIDLMANNDSEDSGCG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1832 MGC117166 protein spire homolog 2 Spir-2 SPIR2 spire homolog 2 (Drosophila); Protein spire homolog 2; Spir-2; spire actin nucleation factor 2; spire family actin nucleation factor 2; spire homolog 2; spire-type actin nucleation factor 2
Gene Aliases: KIAA1832; Spir-2; SPIR2; SPIRE2
UniProt ID: (Human) Q8WWL2
Entrez Gene ID: (Human) 84501
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.