OriGene
Manufacturer Code:TA504591S
Catalog # 50-167-5244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H2 |
Immunogen | Full length human recombit protein of human SPINT1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HAI HAI1 MANSC2; HAI-1; hepatocyte growth factor activator inhibitor 1; Hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; Kunitz-type protease inhibitor 1; serine protease inhibitor, Kunitz type 1
Gene Aliases: HAI; HAI-1; HAI1; MANSC2; SPINT1; UNQ223/PRO256
UniProt ID: (Human) O43278, (Mouse) Q9R097
Entrez Gene ID: (Human) 6692, (Dog) 487501, (Mouse) 20732, (Rat) 311331
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.