Invitrogen
Manufacturer Code:PA537403
Catalog # PIPA537403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length human SPINLW1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 71; cancer/testis antigen 72; CT71; Epididymal protease inhibitor; Eppin; EPPIN EPPIN1 EPPIN2 EPPIN3 WAP7 WFDC7 dJ461P17.2 CT71 CT72 SPINLW1; Protease inhibitor WAP7; serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin); Serine protease inhibitor-like with Kunitz and WAP domains 1; WAP four-disulfide core domain protein 7
Gene Aliases: CT71; CT72; dJ461P17.2; EPPIN; SPINLW1; WAP7; WFDC7
UniProt ID: (Human) O95925
Entrez Gene ID: (Human) 57119
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.