R&D Systems
Manufacturer Code:MAB7496SP
Catalog # MAB7496SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 839305 |
Immunogen | <EM>E. coli</EM>-derived recombinant human SPINK1 Asp24-Cys79 Accession # P00995 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Pancreatic secretory trypsin inhibitor; pancreatic secretory trypsin inhibitor PCTT PCTTSpink3 PSTI PSTISerine protease inhibitor Kazal-type 1 serine peptidase inhibitor Kazal type 1 serine protease inhibitor Kazal type 1 Spink3 TATI TATITumor-associated trypsin inhibitor; Serine protease inhibitor Kazal-type 1; serine protease inhibitor, Kazal type 1; TATI; Tumor-associated trypsin inhibitor
Gene Aliases: PCTT; PSTI; SPINK1; Spink3; TATI; TCP
UniProt ID: (Human) P00995
Entrez Gene ID: (Human) 6690
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.