Proteintech
Manufacturer Code:121051AP150UL
Catalog # 50-172-7717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPIN1 Fusion Protein Ag2750 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 30000 Mr metaphase complex; OCR Ovarian cancer related protein SPIN SPIN1 spindlin 1; Ovarian cancer-related protein; Spindlin-1; Spindlin1; SSEC P
Gene Aliases: OCR; SPIN; SPIN1; TDRD24
UniProt ID: (Human) Q9Y657, (Mouse) Q61142, (Rat) Q4V8J7
Entrez Gene ID: (Human) 10927, (Mouse) 20729, (Rat) 361217
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.