Invitrogen
Manufacturer Code:PA5111601
Catalog # PIPA5111601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human SPHK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C76851 SK 2 SK2 SK-2 Sphingosine kinase 2 Sphk2 SPK 2 SPK-2; SK 2; Sphingosine kinase 2; sphingosine kinase type 2
Gene Aliases: SK 2; SK-2; SK2; SPHK2; SPK 2; SPK-2
UniProt ID: (Human) Q9NRA0
Entrez Gene ID: (Human) 56848
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.