Invitrogen
Manufacturer Code:PA514068
Catalog # PIPA514068
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 286-315 amino acids from the central region of human SPHK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetyltransferase SPHK1; SK 1; Sphingosine kinase 1; Sphingosine kinase 1 SK 1 SPK 1; SPK 1
Gene Aliases: 1110006G24Rik; SK1; SPHK; SPHK1; SPK; Spk1
UniProt ID: (Human) Q9NYA1, (Rat) Q91V26, (Mouse) Q8CI15
Entrez Gene ID: (Human) 8877, (Rat) 170897, (Mouse) 20698
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.