Abnova
Manufacturer Code:PAB9035
Catalog # 89-134-108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 180 of mouse Sphk1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Sphk1.
For Research Use Only
Protein Aliases: 1110006G24Rik; Acetyltransferase SPHK1; SK 1; SK1; Sphingosine kinase 1; SPK 1
Gene Aliases: 1110006G24Rik; Sk1; Sphk1; Spk1
UniProt ID: (Mouse) Q8CI15
Entrez Gene ID: (Mouse) 20698
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.