Novus Biologicals
Manufacturer Code:NBP147203
Catalog # NBP147203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-EASGTDVKQLDQGNK corresponding to internal region according to NP_055902.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0610SPARTIN spastic paraplegia 20 (Troyer syndrome) Spastic paraplegia 20 protein TAHCCP1spartin Trans-activated by hepatitis C virus core protein 1; Spartin; Spastic paraplegia 20 protein; Trans-activated by hepatitis C virus core protein 1
Gene Aliases: KIAA0610; SPART; SPARTIN; SPG20; TAHCCP1
UniProt ID: (Human) Q8N0X7
Entrez Gene ID: (Human) 23111
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.