Invitrogen
Manufacturer Code:PA562301
Catalog # PIPA562301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SPESP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Equatorial segment protein; ESP; Glycosylated 38 kDa sperm protein C-7/8; SP-ESP; Sperm equatorial segment protein 1; Sperm equatorial segment protein 1 ESP Equatorial segment protein SP-ESP Glycosylated 38 kDa sperm protein C-7/8 SPESP1
Gene Aliases: ESP; SP-ESP; SPESP1; UNQ732/PRO1418
UniProt ID: (Human) Q6UW49
Entrez Gene ID: (Human) 246777
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.