Invitrogen
Manufacturer Code:PA561146
Catalog # PIPA561146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SPCS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Microsomal signal peptidase 12 kDa subunit; signal peptidase 12kDa; signal peptidase complex 12 kDa subunit; Signal peptidase complex subunit 1; signal peptidase complex subunit 1 homolog; SPase 12 kDa subunit; SPC1 SPC12 YJR010C-A SPase 12 kDa subunit microsomal signal peptidase 12 kDa subunit signal peptidase 12kDa signal peptidase complex subunit 1
Gene Aliases: HSPC033; SPC1; SPC12; SPCS1; YJR010C-A
UniProt ID: (Human) Q9Y6A9
Entrez Gene ID: (Human) 28972
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.