Invitrogen
Manufacturer Code:PA544731
Catalog # PIPA544731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SPATA7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 296; fertility related protein WMP1; fertility-related protein WMP1; HSD-3.1; sperm DNA no.3; Spermatogenesis-associated protein 7; Spermatogenesis-associated protein 7 homolog; Spermatogenesis-associated protein HSD3
Gene Aliases: AI661438; B230306G18Rik; HEL-S-296; HSD-3.1; HSD3; LCA3; RSD-3; si:ch211-51o12.1; SPATA7; Wmp1
UniProt ID: (Human) Q9P0W8, (Mouse) Q80VP2
Entrez Gene ID: (Bovine) 510352, (Horse) 100061948, (Rabbit) 100351351, (Human) 55812, (Dog) 480411, (Pig) 100153278, (Mouse) 104871, (Rat) 192225, (Guinea pig) 100732301, (Zebrafish) 793830
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.