Invitrogen
Manufacturer Code:PA5100336
Catalog # PIPA5100336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SPATA7 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI661438 B230306G18Rik epididymis secretory protein Li 296 HEL-S-296 HSD3 HSD-3.1 LCA3 SPATA7 spermatogenesis associated 7 spermatogenesis-associated protein 7 Spermatogenesis-associated protein 7 homolog spermatogenesis-associated protein HSD3; epididymis secretory protein Li 296; HSD-3.1; Spermatogenesis-associated protein 7; Spermatogenesis-associated protein 7 homolog; Spermatogenesis-associated protein HSD3
Gene Aliases: AI661438; B230306G18Rik; HEL-S-296; HSD-3.1; HSD3; LCA3; SPATA7
UniProt ID: (Human) Q9P0W8, (Mouse) Q80VP2
Entrez Gene ID: (Human) 55812, (Mouse) 104871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.