Novus Biologicals
Manufacturer Code:NBP274309
Catalog # NBP274309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6H9 |
Immunogen | Full length human recombinant protein of human SPATA22(NP_115987) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: NYDSP20 NYD-SP20 spermatogenesis associated 22 spermatogenesis-associated protein 22 testes development-related NYD-SP20 Testis development protein NYD-SP20; Spermatogenesis-associated protein 22; testicular tissue protein Li 186; Testis development protein NYD-SP20
Gene Aliases: NYD-SP20; NYDSP20; SPATA22
UniProt ID: (Human) Q8NHS9
Entrez Gene ID: (Human) 84690
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.