Invitrogen
Manufacturer Code:PA544923
Catalog # PIPA544923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SPATA22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Spermatogenesis-associated protein 22; testicular tissue protein Li 186; Testis development protein NYD-SP20
Gene Aliases: Gm882; NYD-SP20; NYDSP20; Repro42; RGD1565378; Sgn; SPATA22
UniProt ID: (Bovine) Q2TA20, (Human) Q8NHS9, (Mouse) Q5SV06
Entrez Gene ID: (Dog) 106557466, (Bovine) 539692, (Horse) 100060419, (Rabbit) 100353117, (Human) 84690, (Mouse) 380709, (Rat) 360565, (Guinea pig) 100729635
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.