Invitrogen
Manufacturer Code:PA571864
Catalog # PIPA571864
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 92-120 amino acids from the N-terminal region of human SPATA18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondria-eating protein; Mitochondria-eating protein Spermatogenesis-associated protein 18 SPATA18 MIEAP; spermatogenesis associated 18 homolog; Spermatogenesis-associated protein 18; testis tissue sperm-binding protein Li 71n
Gene Aliases: 1700067I02Rik; MIEAP; SPATA18; SPETEX1
UniProt ID: (Human) Q8TC71, (Mouse) Q0P557
Entrez Gene ID: (Human) 132671, (Mouse) 73472
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.